Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-5-bromo-4-(trifluoromethyl)pyridine, 95%

Click to view available options
Quantity:
250mg
1g
944401-56-3
C6H4BrF3N2
241.011
MFCD09864896
AEWYLVDLWQPJGW-UHFFFAOYSA-N
5-bromo-4-trifluoromethyl pyridin-2-amine,2-amino-5-bromo-4-trifluoromethyl pyridine,5-bromo-4-trifluoromethyl-2-pyridylamine,2-amino-5-bromo-4-trifluoromethylpyridine,acmc-209rru,2-amino-4-trifluoromethyl-5-bromopyridine,2-amino-5-bromo-4-trifluoromethyl pyridine,5-bromo-4-trifluoromethyl-2-pyridinamine,5-bromo-4-trifluoromethyl-2-pyridinylamine,5-bromanyl-4-trifluoromethyl pyridin-2-amine
28875348
5-bromo-4-(trifluoromethyl)pyridin-2-amine
C1=C(C(=CN=C1N)Br)C(F)(F)F
944401-56-3
C6H4BrF3N2
241.011
MFCD09864896
AEWYLVDLWQPJGW-UHFFFAOYSA-N
5-bromo-4-trifluoromethyl pyridin-2-amine,2-amino-5-bromo-4-trifluoromethyl pyridine,5-bromo-4-trifluoromethyl-2-pyridylamine,2-amino-5-bromo-4-trifluoromethylpyridine,acmc-209rru,2-amino-4-trifluoromethyl-5-bromopyridine,2-amino-5-bromo-4-trifluoromethyl pyridine,5-bromo-4-trifluoromethyl-2-pyridinamine,5-bromo-4-trifluoromethyl-2-pyridinylamine,5-bromanyl-4-trifluoromethyl pyridin-2-amine
28875348
5-bromo-4-(trifluoromethyl)pyridin-2-amine
C1=C(C(=CN=C1N)Br)C(F)(F)F
Melting Point 66°C to 75°C
Color Orange to Yellow
Quantity 250mg
Beilstein 15519034
Formula Weight 241.01
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-Amino-5-bromo-4-(trifluoromethyl)pyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning