Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-bromo-6-methyl-4-pyrimidinol, 99%, ACROS Organics™

Click to view available options
Quantity:
50g
Packaging:
Glass Bottle
6307-35-3
C5H6BrN3O
204.027
MFCD00006092
ADLWOFHKMXUDKF-UHFFFAOYSA-N
2-amino-5-bromo-6-methyl-4-pyrimidinol,2-amino-5-bromo-4-hydroxy-6-methylpyrimidine,2-amino-5-bromo-6-methylpyrimidin-4-ol,2-amino-5-bromo-6-methyl-4 3h-pyrimidinone,abmp,pubchem6984,acmc-20ao4q,2-amino-5-bromo-6-methylpyrimidin-4 1h-one,5-bromo-6-methylisocytosine,chembl80800
80567
2-amino-5-bromo-6-methyl-1H-pyrimidin-4-one
CC1=C(C(=O)N=C(N1)N)Br
6307-35-3
C5H6BrN3O
204.027
MFCD00006092
ADLWOFHKMXUDKF-UHFFFAOYSA-N
2-amino-5-bromo-6-methyl-4-pyrimidinol,2-amino-5-bromo-4-hydroxy-6-methylpyrimidine,2-amino-5-bromo-6-methylpyrimidin-4-ol,2-amino-5-bromo-6-methyl-4 3h-pyrimidinone,abmp,pubchem6984,acmc-20ao4q,2-amino-5-bromo-6-methylpyrimidin-4 1h-one,5-bromo-6-methylisocytosine,chembl80800
80567
2-amino-5-bromo-6-methyl-1H-pyrimidin-4-one
CC1=C(C(=O)N=C(N1)N)Br
Packaging Glass Bottle
Quantity 50g
Color White
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 24, 350
Formula Weight 204.03
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material 2-Amino-5-bromo-6-methyl-4-pyrimidinol, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.