Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-5-bromobenzophenone, 97%

Click to view available options
Quantity:
250mg
1g
5g
39859-36-4
C13H10BrNO
276.133
MFCD00059962
LXJVUGANBDAASB-UHFFFAOYSA-N
2-benzoyl-4-bromoaniline,2-amino-5-bromobenzophenone,2-amino-5-bromophenyl phenyl methanone,5-bromo-2-aminobenzophenone,2-amino-5-bromo-phenyl-phenyl-methanone,methanone, 2-amino-5-bromophenyl phenyl,pubchem14551,2-amino-5-bromo benzophenone,2-amino-5-bromo-benzophenone,zerenex e/6030975
99677
(2-amino-5-bromophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)Br)N
39859-36-4
C13H10BrNO
276.133
MFCD00059962
LXJVUGANBDAASB-UHFFFAOYSA-N
2-benzoyl-4-bromoaniline,2-amino-5-bromobenzophenone,2-amino-5-bromophenyl phenyl methanone,5-bromo-2-aminobenzophenone,2-amino-5-bromo-phenyl-phenyl-methanone,methanone, 2-amino-5-bromophenyl phenyl,pubchem14551,2-amino-5-bromo benzophenone,2-amino-5-bromo-benzophenone,zerenex e/6030975
99677
(2-amino-5-bromophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)Br)N
Color Yellow
Melting Point 109°C to 113°C
Quantity 5g
UN Number UN2811
Beilstein 475641
Formula Weight 276.13
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromobenzophenone

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : -30° to -10°C