Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-bromonicotinic acid, 95%, Acros Organics

Click to view available options
Quantity:
1g
52833-94-0
C6H5BrN2O2
217
MFCD01860227
IEPDTLRHISNBLB-UHFFFAOYSA-N
2-amino-5-bromonicotinic acid,2-amino-5-bromo-nicotinic acid,3-pyridinecarboxylic acid, 2-amino-5-bromo,2-amino-5-bromonicotonic acid,2-amino-5-bromo-3-pyridinecarboxylic acid,2-amino-5-bromo-pyridine-3-carboxylic acid,2-amino-5-bromo nicotinic acid,zlchem 461,pubchem5708
737445
2-amino-5-bromopyridine-3-carboxylic acid
C1=C(C(=NC=C1Br)N)C(=O)O
52833-94-0
C6H5BrN2O2
217
MFCD01860227
IEPDTLRHISNBLB-UHFFFAOYSA-N
2-amino-5-bromonicotinic acid,2-amino-5-bromo-nicotinic acid,3-pyridinecarboxylic acid, 2-amino-5-bromo,2-amino-5-bromonicotonic acid,2-amino-5-bromo-3-pyridinecarboxylic acid,2-amino-5-bromo-pyridine-3-carboxylic acid,2-amino-5-bromo nicotinic acid,zlchem 461,pubchem5708
737445
2-amino-5-bromopyridine-3-carboxylic acid
C1=C(C(=NC=C1Br)N)C(=O)O
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 94% min. (HPLC)
Formula Weight 217
Percent Purity 95%
Chemical Name or Material 2-Amino-5-bromonicotinic acid

GHS Signal Word: Warning