Learn More
Alfa Aesar™ 2-Amino-5-bromopyridine, 98%
Description
2-Amino-5-bromopyridine is used for the synthesis of polycyclic azaarenes.
Solubility
Soluble in methanol, chloroform, ethyl acetate. Slightly soluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Light sensitive. Store under argon.
Specifications
Melting Point | 135°C to 140°C |
Quantity | 5g |
Beilstein | 108737 |
Solubility Information | Soluble in methanol,chloroform,ethyl acetate. Slightly soluble in water. |
Formula Weight | 173.01 |
Percent Purity | 98% |
Chemical Name or Material | 2-Amino-5-bromopyridine |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 214-019-9
TSCA : No
Recommended Storage : Ambient temperatures