Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-bromopyrimidine 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
7752-82-1
C4H4BrN3
174.001
UHRHPPKWXSNZLR-UHFFFAOYSA-N
2-amino-5-bromopyrimidine,5-bromopyrimidin-2-ylamine,2-pyrimidinamine, 5-bromo,5-bromo-2-aminopyrimidine,5-bromo-2-pyrimidinamine,pyrimidine, 2-amino-5-bromo,5-bromopyrimidine-2-amine,2-amino-5-bromo-pyrimidine,5-bromopyrimidine-2-ylamine,5-bromo-pyrimidin-2-ylamine
231310
5-bromopyrimidin-2-amine
C1=C(C=NC(=N1)N)Br
7752-82-1
C4H4BrN3
174.001
UHRHPPKWXSNZLR-UHFFFAOYSA-N
2-amino-5-bromopyrimidine,5-bromopyrimidin-2-ylamine,2-pyrimidinamine, 5-bromo,5-bromo-2-aminopyrimidine,5-bromo-2-pyrimidinamine,pyrimidine, 2-amino-5-bromo,5-bromopyrimidine-2-amine,2-amino-5-bromo-pyrimidine,5-bromopyrimidine-2-ylamine,5-bromo-pyrimidin-2-ylamine
231310
5-bromopyrimidin-2-amine
C1=C(C=NC(=N1)N)Br
Color Yellow
Melting Point 244°C
Quantity 25g
UN Number 3077
Formula Weight 174.00
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 2-Amino-5-bromopyrimidine