Alfa Aesar™ 2-Amino-5-chloro-3-fluoropyridine, 98%

Manufacturer: Alfa Aesar™ H30135MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH30135MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 246847-98-3
    Molecular Formula C5H4ClFN2
    Formula Weight 146.55
    MDL Number MFCD09878434
    Synonym 5-Chloro-3-fluoro-2-pyridinamine
    Chemical Name or Material 2-Amino-5-chloro-3-fluoropyridine

    Specifications

    Formula Weight 146.55
    Melting Point 95° to 97°C
    Quantity 250mg
    Molecular Formula C5H4ClFN2
    MDL Number MFCD09878434
    Synonym 5-Chloro-3-fluoro-2-pyridinamine
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    CAS 246847-98-3
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 2-Amino-5-chloro-3-fluoropyridine
    Assay 98%

    GHS H Statement
    H302-H312-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit