Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-5-chlorobenzophenone, 98+%

Click to view available options
Quantity:
100g
500g
2500g
719-59-5
C13H10ClNO
231.679
MFCD00007839
ZUWXHHBROGLWNH-UHFFFAOYSA-N
2-amino-5-chlorobenzophenone,2-amino-5-chlorophenyl phenyl methanone,2-benzoyl-4-chloroaniline,methanone, 2-amino-5-chlorophenyl phenyl,2-amino-5-chlorbenzophenone,5-chloro-2-aminobenzophenone,2-amino-5-chlorobenzylphenone,benzophenone, 2-amino-5-chloro,unii-fr80014zbt,2-amino-5-chlorophenyl phenylmethanone
12870
(2-amino-5-chlorophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)Cl)N
719-59-5
C13H10ClNO
231.679
MFCD00007839
ZUWXHHBROGLWNH-UHFFFAOYSA-N
2-amino-5-chlorobenzophenone,2-amino-5-chlorophenyl phenyl methanone,2-benzoyl-4-chloroaniline,methanone, 2-amino-5-chlorophenyl phenyl,2-amino-5-chlorbenzophenone,5-chloro-2-aminobenzophenone,2-amino-5-chlorobenzylphenone,benzophenone, 2-amino-5-chloro,unii-fr80014zbt,2-amino-5-chlorophenyl phenylmethanone
12870
(2-amino-5-chlorophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)Cl)N
Applications
It is used in the heterocyclic syntheses based on the reactions of dimethyl acetylenedicarboxylate.

Solubility
Solubility in methanol gives very faint turbidity. Soluble in DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Flash Point 211°C (412°F)
Density 1.33
Melting Point 96°C to 99°C
Quantity 2500g
Beilstein 475640
Formula Weight 231.68
Percent Purity ≥98%
Chemical Name or Material 2-Amino-5-chlorobenzophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 211-949-7

RTECSNumber : PC4933500

TSCA : Yes

Recommended Storage : Ambient temperatures