Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-chlorobenzotrifluoride, 97%, ACROS Organics™

Click to view available options
Quantity:
100g
Packaging:
Glass bottle
445-03-4
C7H5ClF3N
195.569
MFCD00007841
CVINWVPRKDIGLL-UHFFFAOYSA-N
2-amino-5-chlorobenzotrifluoride,4-chloro-2-trifluoromethyl aniline,benzenamine, 4-chloro-2-trifluoromethyl,2-amino-5-chlorobenzo trifluoride,5-chloro-2-aminobenzotrifluoride,2-amino-5-chloro-trifluorotoluene,2-trifluoromethyl-4-chloro-aniline,2-trifluoromethyl-4-chloroaniline,4-chloro-alpha,alpha,alpha-trifluoro-o-toluidine,4-chloro-2-trifluoromethyl-phenylamine
67961
4-chloro-2-(trifluoromethyl)aniline
C1=CC(=C(C=C1Cl)C(F)(F)F)N
445-03-4
C7H5ClF3N
195.569
MFCD00007841
CVINWVPRKDIGLL-UHFFFAOYSA-N
2-amino-5-chlorobenzotrifluoride,4-chloro-2-trifluoromethyl aniline,benzenamine, 4-chloro-2-trifluoromethyl,2-amino-5-chlorobenzo trifluoride,5-chloro-2-aminobenzotrifluoride,2-amino-5-chloro-trifluorotoluene,2-trifluoromethyl-4-chloro-aniline,2-trifluoromethyl-4-chloroaniline,4-chloro-alpha,alpha,alpha-trifluoro-o-toluidine,4-chloro-2-trifluoromethyl-phenylamine
67961
4-chloro-2-(trifluoromethyl)aniline
C1=CC(=C(C=C1Cl)C(F)(F)F)N
Quantity 100g
Infrared Spectrum Authentic
Color Yellow
Packaging Glass bottle
Refractive Index 1.5059 to 1.5079 (20°C, 589nm)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula H2NC6H3(Cl)CF3
Beilstein 12, III, 1921
Formula Weight 195.57
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 2-Amino-5-chlorobenzotrifluoride, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger