Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-5-chlorophenol, ≥97%

Click to view available options
Quantity:
25g
100g
28443-50-7
C6H6ClNO
143.57
MFCD02093863
FZCQMIRJCGWWCL-UHFFFAOYSA-N
4-chloro-2-hydroxyaniline,phenol, 2-amino-5-chloro,2-amino-5-chloro-phenol,5-chloro-2-aminophenol,pubchem1478,5-chloro-o-aminophenol,5-chloro-2-amino phenol,2-hydroxy-4-chloroaniline,acmc-209h2j,2-azanyl-5-chloranyl-phenol
91591
CHEBI:75051
2-amino-5-chlorophenol
C1=CC(=C(C=C1Cl)O)N
28443-50-7
C6H6ClNO
143.57
MFCD02093863
FZCQMIRJCGWWCL-UHFFFAOYSA-N
4-chloro-2-hydroxyaniline,phenol, 2-amino-5-chloro,2-amino-5-chloro-phenol,5-chloro-2-aminophenol,pubchem1478,5-chloro-o-aminophenol,5-chloro-2-amino phenol,2-hydroxy-4-chloroaniline,acmc-209h2j,2-azanyl-5-chloranyl-phenol
91591
CHEBI:75051
2-amino-5-chlorophenol
C1=CC(=C(C=C1Cl)O)N
Melting Point 145°C to 153°C
Quantity 100g
Color Brown
UN Number UN2811
Beilstein 386421
Formula Weight 143.57
Physical Form Powder
Percent Purity ≥97%
Chemical Name or Material 2-Amino-5-chlorophenol

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA