Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-5-chloropyrazine, 95%

Click to view available options
Quantity:
1g
5g
25g
33332-29-5
C4H4ClN3
129.547
MFCD03423596
HWCKAMAVEWVBDO-UHFFFAOYSA-N
2-amino-5-chloropyrazine,pyrazinamine, 5-chloro,5-amino-2-chloropyrazine,5-chloro-pyrazin-2-ylamine,2-amino-5-chloro-1,4-diazine,pubchem17501,acmc-1cix9,5-chloro-2-amino-pyrazine,5-chloro-2-pyrazinamine,3-amino-6-chloropyrazine
12808419
5-chloropyrazin-2-amine
C1=C(N=CC(=N1)Cl)N
33332-29-5
C4H4ClN3
129.547
MFCD03423596
HWCKAMAVEWVBDO-UHFFFAOYSA-N
2-amino-5-chloropyrazine,pyrazinamine, 5-chloro,5-amino-2-chloropyrazine,5-chloro-pyrazin-2-ylamine,2-amino-5-chloro-1,4-diazine,pubchem17501,acmc-1cix9,5-chloro-2-amino-pyrazine,5-chloro-2-pyrazinamine,3-amino-6-chloropyrazine
12808419
5-chloropyrazin-2-amine
C1=C(N=CC(=N1)Cl)N
Quantity 1g
Melting Point 122°C to 128°C
Beilstein 507879
Formula Weight 129.55
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-Amino-5-chloropyrazine

storageNote1 : 2° to 8°C