Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-ethyl-1,3,4-thiadiazole, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
14068-53-2
C4H7N3S
129.181
MFCD00003111
QXTRPGAMVIONMK-UHFFFAOYSA-N
2-amino-5-ethyl-1,3,4-thiadiazole,1,3,4-thiadiazol-2-amine, 5-ethyl,1,3,4-thiadiazole, 2-amino-5-ethyl,5-ethyl-1,3,4 thiadiazol-2-ylamine,5-ethyl-1,3,4-thiadiazole-2-ylamine,acmc-209wwz,4-27-00-08073 beilstein handbook reference,aronis24978,tos-bb-1085,1,4-thiadiazol-2-amine, 5-ethyl
26444
CHEBI:86540
5-ethyl-1,3,4-thiadiazol-2-amine
CCC1=NN=C(S1)N
14068-53-2
C4H7N3S
129.181
MFCD00003111
QXTRPGAMVIONMK-UHFFFAOYSA-N
2-amino-5-ethyl-1,3,4-thiadiazole,1,3,4-thiadiazol-2-amine, 5-ethyl,1,3,4-thiadiazole, 2-amino-5-ethyl,5-ethyl-1,3,4 thiadiazol-2-ylamine,5-ethyl-1,3,4-thiadiazole-2-ylamine,acmc-209wwz,4-27-00-08073 beilstein handbook reference,aronis24978,tos-bb-1085,1,4-thiadiazol-2-amine, 5-ethyl
26444
CHEBI:86540
5-ethyl-1,3,4-thiadiazol-2-amine
CCC1=NN=C(S1)N
Color Beige
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Formula Weight 129.19
Physical Form Crystalline Powder
Percent Purity ≥98.0%
Chemical Name or Material 2-Amino-5-ethyl-1, 3, 4-thiadiazole, 98+%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 237-921-4

RTECSNumber : XI3095000

TSCA : TSCA