Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-5-fluoro-6-methylpyridine, 98%

Click to view available options
Quantity:
1g
5g
110919-71-6
C6H7FN2
126.134
MFCD09835298
WMGRXNKQEVOVET-UHFFFAOYSA-N
2-amino-5-fluoro-6-methylpyridine,5-fluoro-6-methyl-2-pyridinamine,2-pyridinamine, 5-fluoro-6-methyl,2-amino-5-fluoro-6-methyl-pyridine,5-fluoro-6-methyl-pyridin-2-ylamine,5-fluoro-6-methyl-2-pyridylamine,6-amino-3-fluoro-2-methylpyridine
13789841
5-fluoro-6-methylpyridin-2-amine
CC1=C(C=CC(=N1)N)F
110919-71-6
C6H7FN2
126.134
MFCD09835298
WMGRXNKQEVOVET-UHFFFAOYSA-N
2-amino-5-fluoro-6-methylpyridine,5-fluoro-6-methyl-2-pyridinamine,2-pyridinamine, 5-fluoro-6-methyl,2-amino-5-fluoro-6-methyl-pyridine,5-fluoro-6-methyl-pyridin-2-ylamine,5-fluoro-6-methyl-2-pyridylamine,6-amino-3-fluoro-2-methylpyridine
13789841
5-fluoro-6-methylpyridin-2-amine
CC1=C(C=CC(=N1)N)F
Quantity 5g
Color White
UN Number UN2811
Beilstein 4376910
Formula Weight 126.13
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Amino-5-fluoro-6-methylpyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.