Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-5-fluorobenzothiazole, 98%

Manufacturer:  Alfa Aesar™ H51022MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51022MD


  Check Availability
Add to cart

Specifications

Description

Applications
2-Amino-5-fluorobenzothiazole is used as organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

20358-07-0
168.19
5-Fluoro-2-benzothiazolamine
C7H5FN2S
MFCD05664560
2-Amino-5-fluorobenzothiazole

Specifications

20358-07-0
168.19
250mg
2-Amino-5-fluorobenzothiazole
C7H5FN2S
MFCD05664560
5-Fluoro-2-benzothiazolamine
98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit