Alfa Aesar™ 2-Amino-5-fluorophenol, 97%

Manufacturer: Alfa Aesar™ H3292803

Quantity 1g

 View more versions of this product

Catalog No. AAH3292803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 53981-24-1
    Molecular Formula C6H6FNO
    Formula Weight 127.12
    MDL Number MFCD00671759
    Synonym 2-Hydroxy-4-fluoroaniline
    Chemical Name or Material 2-Amino-5-fluorophenol

    Specifications

    Formula Weight 127.12
    Molecular Formula C6H6FNO
    Melting Point 135° to 140°C
    Quantity 1g
    MDL Number MFCD00671759
    CAS 53981-24-1
    Synonym 2-Hydroxy-4-fluoroaniline
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P312
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Recommended Storage 2° to 8°C
    Chemical Name or Material 2-Amino-5-fluorophenol
    Assay 97%

    Description

    Applications
    2-Amino-5-fluorophenol is a fluoroaniline metabolite. 2-Amino-5-fluorophenol is used as organic intermediate. To synthesize the intermediate (2-amino-5-fluorophenol-N, N, O-triacetic acid trimethyl ester) of indicator which used in the determination Mg2+ by NMR, three feasible reactions were experimentally carried out, such as alkaline hydrolysis, catalytic hydrogenation and esterification.

    Solubility
    Slightly soluble in water.

    Notes
    Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerated.

    GHS H Statement
    H302-H312-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P280h-P305+P351+P338-P312
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Call a POISON CENTER or doctor/physician if you feel unwell.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit