Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-iodopyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
20511-12-0
C5H5IN2
220.013
MFCD00160312
IVILGUFRMDBUEQ-UHFFFAOYSA-N
2-amino-5-iodopyridine,2-pyridinamine, 5-iodo,5-iodo-pyridin-2-ylamine,5-iodo-2-pyridinamine,5-iodo-2-aminopyridine,2-amino-5-iodo pyridine,pyridine, 2-amino-5-iodo,5-iod-2-pyridinamin,5-iodo-2-pyridylamine,6-amino-3-iodopyridine
88579
5-iodopyridin-2-amine
C1=CC(=NC=C1I)N
20511-12-0
C5H5IN2
220.013
MFCD00160312
IVILGUFRMDBUEQ-UHFFFAOYSA-N
2-amino-5-iodopyridine,2-pyridinamine, 5-iodo,5-iodo-pyridin-2-ylamine,5-iodo-2-pyridinamine,5-iodo-2-aminopyridine,2-amino-5-iodo pyridine,pyridine, 2-amino-5-iodo,5-iod-2-pyridinamin,5-iodo-2-pyridylamine,6-amino-3-iodopyridine
88579
5-iodopyridin-2-amine
C1=CC(=NC=C1I)N
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
Color Beige to Yellow
Melting Point 127°C to 131°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 220.01
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Amino-5-iodopyridine, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.