Alfa Aesar™ 2-Amino-5-methylbenzenesulfonic acid, 99%

Manufacturer: Alfa Aesar™ B2456018

Quantity 50g

 View more versions of this product

Catalog No. AAB2456018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 88-44-8
    Molecular Formula C7H9NO3S
    Formula Weight 187.22
    MDL Number MFCD00007908
    UN Number UN3261
    Beilstein 2211509
    Synonym 4-Aminotoluene-3-sulfonic acid; p-Toluidine-3-sulfonic acid
    Chemical Name or Material 2-Amino-5-methylbenzenesulfonic acid

    Specifications

    Formula Weight 187.22
    UN Number UN3261
    Quantity 50g
    Health Hazard 1 Danger
    RTECS Number XT6320000
    TSCA TSCA
    Molecular Formula C7H9NO3S
    MDL Number MFCD00007908
    CAS 88-44-8
    Beilstein 2211509
    Synonym 4-Aminotoluene-3-sulfonic acid; p-Toluidine-3-sulfonic acid
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 2-Amino-5-methylbenzenesulfonic acid
    Assay 99%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit