Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-5-methylbenzonitrile, 97%

Click to view available options
Quantity:
250mg
1g
5g
5925-93-9
C8H8N2
132.166
MFCD00267506
OZLMBXPYRDASTP-UHFFFAOYSA-N
2-amino-5-methyl-benzonitrile,benzonitrile, 2-amino-5-methyl,2-amino-5-methylbenzonitril,6-amino-m-tolunitrile,pubchem17447,2-cyano-4-methylaniline,acmc-209ww6,2-amino-5-methyl benzonitrile,benzonitrile,2-amino-5-methyl,2-amino-5-methylbenzonitrile
242778
2-amino-5-methylbenzonitrile
CC1=CC(=C(C=C1)N)C#N
5925-93-9
C8H8N2
132.166
MFCD00267506
OZLMBXPYRDASTP-UHFFFAOYSA-N
2-amino-5-methyl-benzonitrile,benzonitrile, 2-amino-5-methyl,2-amino-5-methylbenzonitril,6-amino-m-tolunitrile,pubchem17447,2-cyano-4-methylaniline,acmc-209ww6,2-amino-5-methyl benzonitrile,benzonitrile,2-amino-5-methyl,2-amino-5-methylbenzonitrile
242778
2-amino-5-methylbenzonitrile
CC1=CC(=C(C=C1)N)C#N
Melting Point 59°C to 63°C
Quantity 250mg
UN Number UN3439
Beilstein 1862343
Formula Weight 132.17
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Amino-5-methylbenzonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

storageNote1 : 2° to 8°C