Alfa Aesar™ 2-Amino-5-nitrobenzoic acid, 98%

Manufacturer: Alfa Aesar™ A1042630

Quantity 250g

 View more versions of this product

Catalog No. AAA1042630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 616-79-5
    Molecular Formula C7H6N2O4
    Formula Weight 182.14
    MDL Number MFCD00017039
    Beilstein 646219
    Synonym 5-Nitroanthranilic acid
    Chemical Name or Material 2-Amino-5-nitrobenzoic acid

    Specifications

    Formula Weight 182.14
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 616-79-5
    Beilstein 646219
    Synonym 5-Nitroanthranilic acid
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    Quantity 250g
    Molecular Formula C7H6N2O4
    MDL Number MFCD00017039
    Health Hazard 1 Warning
    Melting Point ≈270°C (dec.)
    Chemical Name or Material 2-Amino-5-nitrobenzoic acid
    Assay 98%

    Description

    Applications
    2-Amino-5-nitrobenzoic acid, is used as an intermediate for dyes and pharmaceuticals.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatabile to strong oxidizing agents. Stable under ordinary conditions.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit