Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-nitrobenzotrifluoride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
121-01-7
C7H5F3N2O2
206.124
MFCD00007365
HOTZLWVITTVZGY-UHFFFAOYSA-N
2-amino-5-nitrobenzotrifluoride,4-nitro-2-trifluoromethyl aniline,2-amino-5-nitro-trifluorobenzene,4-nitro-alpha,alpha,alpha-trifluoro-o-toluidine,4-nitro-2-trifluoromethyl benzenamine,4-nitro-a,a,a-trifluoro-o-toluidine,benzenamine, 4-nitro-2-trifluoromethyl,5-nitro-2-aminobenzotrifluoride,2-trifluoromethyl-4-nitroaniline,a,a,a-trifluoro-4-nitro-o-toluidine
67128
4-nitro-2-(trifluoromethyl)aniline
C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)N
121-01-7
C7H5F3N2O2
206.124
MFCD00007365
HOTZLWVITTVZGY-UHFFFAOYSA-N
2-amino-5-nitrobenzotrifluoride,4-nitro-2-trifluoromethyl aniline,2-amino-5-nitro-trifluorobenzene,4-nitro-alpha,alpha,alpha-trifluoro-o-toluidine,4-nitro-2-trifluoromethyl benzenamine,4-nitro-a,a,a-trifluoro-o-toluidine,benzenamine, 4-nitro-2-trifluoromethyl,5-nitro-2-aminobenzotrifluoride,2-trifluoromethyl-4-nitroaniline,a,a,a-trifluoro-4-nitro-o-toluidine
67128
4-nitro-2-(trifluoromethyl)aniline
C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)N
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow to Brown
Quantity 25g
Melting Point 90°C to 93°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula H2NC6H3(NO2)CF3
Formula Weight 206.12
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Amino-5-nitrobenzotrifluoride, 98%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-443-2

TSCA : TSCA