Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-nitrothiazole 98.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
121-66-4
C3H3N3O2S
145.136
MFCD00005326
MIHADVKEHAFNPG-UHFFFAOYSA-N
2-amino-5-nitrothiazole,5-nitrothiazol-2-amine,enheptin,entramin,2-thiazolamine, 5-nitro,nitramin ido,amnizol soluble,enheptin-t,5-nitro-2-thiazolamine,enheptin premix
8486
CHEBI:82386
5-nitro-1,3-thiazol-2-amine
C1=C(SC(=N1)N)[N+](=O)[O-]
121-66-4
C3H3N3O2S
145.136
MFCD00005326
MIHADVKEHAFNPG-UHFFFAOYSA-N
2-amino-5-nitrothiazole,5-nitrothiazol-2-amine,enheptin,entramin,2-thiazolamine, 5-nitro,nitramin ido,amnizol soluble,enheptin-t,5-nitro-2-thiazolamine,enheptin premix
8486
CHEBI:82386
5-nitro-1,3-thiazol-2-amine
C1=C(SC(=N1)N)[N+](=O)[O-]
Color Undesignated
Quantity 500g
Melting Point 202°C
Formula Weight 145.14
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material 2-Amino-5-nitrothiazole

EINECSNumber : (5)-0206

RTECSNumber : XJ2800000

TSCA : Yes