Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-phenylpyrazine, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
13535-13-2
C10H9N3
171.2
KJAKXVBZQBPPOB-UHFFFAOYSA-N
2-amino-5-phenylpyrazine,pyrazinamine, 5-phenyl,pyrazine, 2-amino-5-phenyl,acmc-1bpgx,5-phenyl-2-pyrazinamine,5-phenyl-2-aminopyrazine,2-pyrazinamine,5-phenyl,5-phenylpyrazine-2-ylamine,5-phenyl-2-pyrazinamine #
598734
5-phenylpyrazin-2-amine
C1=CC=C(C=C1)C2=CN=C(C=N2)N
13535-13-2
C10H9N3
171.2
KJAKXVBZQBPPOB-UHFFFAOYSA-N
2-amino-5-phenylpyrazine,pyrazinamine, 5-phenyl,pyrazine, 2-amino-5-phenyl,acmc-1bpgx,5-phenyl-2-pyrazinamine,5-phenyl-2-aminopyrazine,2-pyrazinamine,5-phenyl,5-phenylpyrazine-2-ylamine,5-phenyl-2-pyrazinamine #
598734
5-phenylpyrazin-2-amine
C1=CC=C(C=C1)C2=CN=C(C=N2)N
Packaging Glass Bottle
Quantity 1g
Melting Point 142.0°C to 148.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 171.2
Percent Purity 98%
Chemical Name or Material 2-Amino-5-phenylpyrazine

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritatio

GHS P Statement
Wear eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water f

GHS Signal Word: Warning