Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-trifluoromethyl-1,3,4-thiadiazole, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
10444-89-0
C3H2F3N3S
169.125
MFCD00003109
LTEUXHSAYOSFGQ-UHFFFAOYSA-N
5-trifluoromethyl-1,3,4-thiadiazol-2-amine,2-amino-5-trifluoromethyl-1,3,4-thiadiazole,1,3,4-thiadiazol-2-amine, 5-trifluoromethyl,5-trifluoromethyl-1,3,4 thiadiazol-2-ylamine,1,3,4-thiadiazole, 2-amino-5-trifluoromethyl,5-trifluoromethyl-1,3,4-thiadiazole-2-ylamine,zlchem 1059,pubchem14503,maybridge1_003448
82634
5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine
C1(=NN=C(S1)N)C(F)(F)F
10444-89-0
C3H2F3N3S
169.125
MFCD00003109
LTEUXHSAYOSFGQ-UHFFFAOYSA-N
5-trifluoromethyl-1,3,4-thiadiazol-2-amine,2-amino-5-trifluoromethyl-1,3,4-thiadiazole,1,3,4-thiadiazol-2-amine, 5-trifluoromethyl,5-trifluoromethyl-1,3,4 thiadiazol-2-ylamine,1,3,4-thiadiazole, 2-amino-5-trifluoromethyl,5-trifluoromethyl-1,3,4-thiadiazole-2-ylamine,zlchem 1059,pubchem14503,maybridge1_003448
82634
5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine
C1(=NN=C(S1)N)C(F)(F)F
Packaging Glass bottle
Melting Point 220°C to 225°C
Infrared Spectrum Authentic
Quantity 5g
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 169.13
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Amino-5-trifluoromethyl-1, 3, 4-thiadiazole, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.