Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-5-trifluoromethyl-1,3,4-thiadiazole, 98%

Click to view available options
Quantity:
1g
5g
25g
10444-89-0
C3H2F3N3S
169.125
MFCD00003109
LTEUXHSAYOSFGQ-UHFFFAOYSA-N
5-trifluoromethyl-1,3,4-thiadiazol-2-amine,2-amino-5-trifluoromethyl-1,3,4-thiadiazole,1,3,4-thiadiazol-2-amine, 5-trifluoromethyl,5-trifluoromethyl-1,3,4 thiadiazol-2-ylamine,1,3,4-thiadiazole, 2-amino-5-trifluoromethyl,5-trifluoromethyl-1,3,4-thiadiazole-2-ylamine,zlchem 1059,pubchem14503,maybridge1_003448
82634
5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine
C1(=NN=C(S1)N)C(F)(F)F
10444-89-0
C3H2F3N3S
169.125
MFCD00003109
LTEUXHSAYOSFGQ-UHFFFAOYSA-N
5-trifluoromethyl-1,3,4-thiadiazol-2-amine,2-amino-5-trifluoromethyl-1,3,4-thiadiazole,1,3,4-thiadiazol-2-amine, 5-trifluoromethyl,5-trifluoromethyl-1,3,4 thiadiazol-2-ylamine,1,3,4-thiadiazole, 2-amino-5-trifluoromethyl,5-trifluoromethyl-1,3,4-thiadiazole-2-ylamine,zlchem 1059,pubchem14503,maybridge1_003448
82634
5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine
C1(=NN=C(S1)N)C(F)(F)F
Quantity 5g
Melting Point 222°C to 227°C
Beilstein 513140
Formula Weight 169.13
Percent Purity 98%
Chemical Name or Material 2-Amino-5-trifluoromethyl-1,3,4-thiadiazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 233-930-2

TSCA : Yes

Recommended Storage : Ambient temperatures