Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-6-bromopurine, 98%

Click to view available options
Quantity:
1g
5g
25g
82499-03-4
C5H4BrN5
214.026
MFCD01321309
HPGBGNVPUMCKPM-UHFFFAOYSA-N
2-amino-6-bromopurine,6-bromo-9h-purin-2-amine,1h-purin-2-amine, 6-bromo,6-bromoguanine,d2-amino-6-bromopurine,6-bromopurine-2-ylamine,purine, 2-amino-6-bromo,6-bromo-9h-purin-2-ylamine,9h-purin-2-amine,6-bromo
6101139
6-bromo-7H-purin-2-amine
C1=NC2=C(N1)C(=NC(=N2)N)Br
82499-03-4
C5H4BrN5
214.026
MFCD01321309
HPGBGNVPUMCKPM-UHFFFAOYSA-N
2-amino-6-bromopurine,6-bromo-9h-purin-2-amine,1h-purin-2-amine, 6-bromo,6-bromoguanine,d2-amino-6-bromopurine,6-bromopurine-2-ylamine,purine, 2-amino-6-bromo,6-bromo-9h-purin-2-ylamine,9h-purin-2-amine,6-bromo
6101139
6-bromo-7H-purin-2-amine
C1=NC2=C(N1)C(=NC(=N2)N)Br
Quantity 25g
Melting Point >300°C
Formula Weight 214.02
Percent Purity 98%
Chemical Name or Material 2-Amino-6-bromopurine