Alfa Aesar™ 2-Amino-6-bromopyridine, 98%

Manufacturer: Alfa Aesar™ H2655514

Quantity 25g

 View more versions of this product

Catalog No. AAH2655514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19798-81-3
    Molecular Formula C5H5BrN2
    Formula Weight 173.01
    MDL Number MFCD00137843
    Synonym 6-Bromo-2-pyridinamine
    Chemical Name or Material 2-Amino-6-bromopyridine

    Specifications

    Formula Weight 173.01
    Melting Point 88° to 91°C
    Molecular Formula C5H5BrN2
    MDL Number MFCD00137843
    Synonym 6-Bromo-2-pyridinamine
    Health Hazard 1 Warning
    Quantity 25g
    CAS 19798-81-3
    Health Hazard 2 GHS H Statement
    H302-H312-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2-Amino-6-bromopyridine
    Assay 98%

    Description

    Applications
    Employed in an efficient one-pot synthesis of 7-azaindoles. Used in the synthesis of anti-HIV agents.

    Solubility
    Slightly soluble in water.

    Notes
    Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H302-H312-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit