Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-chloro-3-nitropyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
27048-04-0
C5H4ClN3O2
173.56
WERABQRUGJIMKQ-UHFFFAOYSA-N
2-amino-6-chloro-3-nitropyridine,2-amino-3-nitro-6-chloropyridine,6-chloro-3-nitro-2-pyridylamine,6-chloro-3-nitro-2-pyridinamine,6-amino-2-chloro-5-nitropyridine,6-chloro-3-nitropyridin-2-ylamine,6-chloro-3-nitro-pyridin-2-ylamine,2-pyridinamine, 6-chloro-3-nitro,6-chloro-3-nitro-pyridin-2-amine,zlchem 113
2724035
6-chloro-3-nitropyridin-2-amine
C1=CC(=NC(=C1[N+](=O)[O-])N)Cl
27048-04-0
C5H4ClN3O2
173.56
WERABQRUGJIMKQ-UHFFFAOYSA-N
2-amino-6-chloro-3-nitropyridine,2-amino-3-nitro-6-chloropyridine,6-chloro-3-nitro-2-pyridylamine,6-chloro-3-nitro-2-pyridinamine,6-amino-2-chloro-5-nitropyridine,6-chloro-3-nitropyridin-2-ylamine,6-chloro-3-nitro-pyridin-2-ylamine,2-pyridinamine, 6-chloro-3-nitro,6-chloro-3-nitro-pyridin-2-amine,zlchem 113
2724035
6-chloro-3-nitropyridin-2-amine
C1=CC(=NC(=C1[N+](=O)[O-])N)Cl
Infrared Spectrum Authentic
Quantity 1g
Melting Point 193.0°C to 198.0°C
Packaging Glass Bottle
Assay Percent Range 96% min. (HPLC)
Formula Weight 173.56
Percent Purity 97%
Chemical Name or Material 2-Amino-6-chloro-3-nitropyridine

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 248-188-5