Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-6-chloro-3-nitropyridine, 98%

Click to view available options
Quantity:
25g
100g
27048-04-0
C5H4ClN3O2
173.556
MFCD00044350
WERABQRUGJIMKQ-UHFFFAOYSA-N
2-amino-6-chloro-3-nitropyridine,2-amino-3-nitro-6-chloropyridine,6-chloro-3-nitro-2-pyridylamine,6-chloro-3-nitro-2-pyridinamine,6-amino-2-chloro-5-nitropyridine,6-chloro-3-nitropyridin-2-ylamine,6-chloro-3-nitro-pyridin-2-ylamine,2-pyridinamine, 6-chloro-3-nitro,6-chloro-3-nitro-pyridin-2-amine,zlchem 113
2724035
6-chloro-3-nitropyridin-2-amine
C1=CC(=NC(=C1[N+](=O)[O-])N)Cl
27048-04-0
C5H4ClN3O2
173.556
MFCD00044350
WERABQRUGJIMKQ-UHFFFAOYSA-N
2-amino-6-chloro-3-nitropyridine,2-amino-3-nitro-6-chloropyridine,6-chloro-3-nitro-2-pyridylamine,6-chloro-3-nitro-2-pyridinamine,6-amino-2-chloro-5-nitropyridine,6-chloro-3-nitropyridin-2-ylamine,6-chloro-3-nitro-pyridin-2-ylamine,2-pyridinamine, 6-chloro-3-nitro,6-chloro-3-nitro-pyridin-2-amine,zlchem 113
2724035
6-chloro-3-nitropyridin-2-amine
C1=CC(=NC(=C1[N+](=O)[O-])N)Cl
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water. Soluble in dimethyl sulfoxide and chloroform.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Quantity 100g
Color Yellow
Melting Point 192°C to 196°C
UN Number UN2811
Beilstein 474607
Formula Weight 173.56
Physical Form Crystalline
Percent Purity 98%
Chemical Name or Material 2-Amino-6-chloro-3-nitropyridine

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : 2° to 8°C