Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2-Amino-6-chloro-3-nitropyridine, 98%
Click to view available options
Quantity:
25g
100g
Description
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water. Soluble in dimethyl sulfoxide and chloroform.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water. Soluble in dimethyl sulfoxide and chloroform.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
Color | Yellow |
Melting Point | 192°C to 196°C |
Quantity | 25g |
UN Number | UN2811 |
Beilstein | 474607 |
Solubility Information | Slightly soluble in water. Soluble in dimethyl sulfoxide and chloroform. |
Formula Weight | 173.56 |
Physical Form | Crystalline |
Percent Purity | 98% |
Chemical Name or Material | 2-Amino-6-chloro-3-nitropyridine |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 248-188-5
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold