Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-6-chloro-4-nitrophenol, 97%

Click to view available options
Quantity:
1g
5g
25g
6358-08-3
C6H5ClN2O3
188.57
MFCD00035767
MHAFRUMLQZZSIN-UHFFFAOYSA-N
unii-13d00yq1u0,6-amino-4-chloro-2-nitrophenol,2-amino-4-chloro-6-nitro-phenol,2-amino-4-chloro-6-nitro phenol,phenol, 2-amino-4-chloro-6-nitro,acmc-2097jj,4-phenylethyl isothiocyanate,4-chlor-6-nitro-2-aminophenol,5-chloro-2-hydroxy-3-nitroaniline,phenol,2-amino-4-chloro-6-nitro
4348294
2-amino-4-chloro-6-nitrophenol
C1=C(C=C(C(=C1[N+](=O)[O-])O)N)Cl
6358-08-3
C6H5ClN2O3
188.57
MFCD00035767
MHAFRUMLQZZSIN-UHFFFAOYSA-N
unii-13d00yq1u0,6-amino-4-chloro-2-nitrophenol,2-amino-4-chloro-6-nitro-phenol,2-amino-4-chloro-6-nitro phenol,phenol, 2-amino-4-chloro-6-nitro,acmc-2097jj,4-phenylethyl isothiocyanate,4-chlor-6-nitro-2-aminophenol,5-chloro-2-hydroxy-3-nitroaniline,phenol,2-amino-4-chloro-6-nitro
4348294
2-amino-4-chloro-6-nitrophenol
C1=C(C=C(C(=C1[N+](=O)[O-])O)N)Cl
Melting Point 160°C to 164°C
Quantity 5g
Color Orange
UN Number UN2811
Beilstein 2727724
Formula Weight 188.57
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2-Amino-6-chloro-4-nitrophenol

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : ST5743000

TSCA : TSCA

storageNote1 : -30° to -10°C