Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-6-chloro-4-nitrophenol, 97%

Click to view available options
Quantity:
1g
5g
25g
6373-11-1
C16H8O2
232.24
MFCD00035767
YAIBDWAANBTYIA-UHFFFAOYSA-N
1,2-aceanthrylenedione,aceanthrenequinone,1,2-aceanthrenedione,acenanthrene-9,10-dione,aceanthrenchinon,aceanthrenequinone,acmc-20ap73,d0k6po,1,2-dione-based compound, 16,1,2-dihydroaceanthrylene-1,2-dione
316765
aceanthrylene-1,2-dione
C1=CC=C2C(=C1)C=C3C=CC=C4C3=C2C(=O)C4=O
6373-11-1
C16H8O2
232.24
MFCD00035767
YAIBDWAANBTYIA-UHFFFAOYSA-N
1,2-aceanthrylenedione,aceanthrenequinone,1,2-aceanthrenedione,acenanthrene-9,10-dione,aceanthrenchinon,aceanthrenequinone,acmc-20ap73,d0k6po,1,2-dione-based compound, 16,1,2-dihydroaceanthrylene-1,2-dione
316765
aceanthrylene-1,2-dione
C1=CC=C2C(=C1)C=C3C=CC=C4C3=C2C(=O)C4=O
Melting Point 160°C to 164°C
Quantity 1g
Color Orange
UN Number UN2811
Beilstein 2727724
Formula Weight 188.57
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2-Amino-6-chloro-4-nitrophenol

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : ST5743000

TSCA : TSCA

storageNote1 : -30° to -10°C