Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-chloropurine, 99+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
10310-21-1
C5H4ClN5
169.57
MFCD00075252
RYYIULNRIVUMTQ-UHFFFAOYSA-N
2-amino-6-chloropurine,6-chloroguanine,6-chloro-9h-purin-2-amine,6-chloro-2-aminopurine,2-amino-6-chlorpurine,1h-purin-2-amine, 6-chloro,purine, 2-amino-6-chloro,6-chloro-7h-purin-2-ylamine,2-amino-6-chloro-purin,2-amino-6-chloro-purine
5360349
CHEBI:72345
6-chloro-7H-purin-2-amine
C1=NC2=C(N1)C(=NC(=N2)N)Cl
10310-21-1
C5H4ClN5
169.57
MFCD00075252
RYYIULNRIVUMTQ-UHFFFAOYSA-N
2-amino-6-chloropurine,6-chloroguanine,6-chloro-9h-purin-2-amine,6-chloro-2-aminopurine,2-amino-6-chlorpurine,1h-purin-2-amine, 6-chloro,purine, 2-amino-6-chloro,6-chloro-7h-purin-2-ylamine,2-amino-6-chloro-purin,2-amino-6-chloro-purine
5360349
CHEBI:72345
6-chloro-7H-purin-2-amine
C1=NC2=C(N1)C(=NC(=N2)N)Cl
Melting Point >300°C
Water 0.5% max. (K.F.)
Color White to Yellow
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Solubility Information Solubility in water: 1.7g/L water (20°C)
Formula Weight 169.57
Physical Form Powder
Percent Purity ≥99.0% (on dried substance)
Chemical Name or Material 2-Amino-6-chloropurine, 99+%
Show More Show Less

EINECSNumber : 233-686-7

RTECSNumber : UO7502000

TSCA : TSCA