Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-6-ethylpyridine, 97%

Click to view available options
Quantity:
2.5g
10g
50g
21717-29-3
C7H10N2
122.171
MFCD00218265
JXKAUUVMXZIJNZ-UHFFFAOYSA-N
2-amino-6-ethylpyridine,6-ethyl-2-pyridinamine,2-pyridinamine, 6-ethyl,6-ethyl-2-pyridylamine,acmc-1clku,2-pyridinamine,6-ethyl,2-amino-6-ethyl-pyridine,6-ethyl-pyridin-2-ylamine
89021
6-ethylpyridin-2-amine
CCC1=NC(=CC=C1)N
21717-29-3
C7H10N2
122.171
MFCD00218265
JXKAUUVMXZIJNZ-UHFFFAOYSA-N
2-amino-6-ethylpyridine,6-ethyl-2-pyridinamine,2-pyridinamine, 6-ethyl,6-ethyl-2-pyridylamine,acmc-1clku,2-pyridinamine,6-ethyl,2-amino-6-ethyl-pyridine,6-ethyl-pyridin-2-ylamine
89021
6-ethylpyridin-2-amine
CCC1=NC(=CC=C1)N
Applications
The action of iodine on 2-amino-6-ethylpyridine in an alkaline medium gives 2-amino-5-iodo-6-ethylpyridine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Odor Unpleasant
Quantity 2.5g
UN Number UN2810
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 2-Amino-6-ethylpyridine

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.