Alfa Aesar™ 2-Amino-6-fluorobenzonitrile, 99%

Manufacturer: Alfa Aesar™ A1245606

Quantity 5g

 View more versions of this product

Catalog No. AAA1245606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 77326-36-4
    Molecular Formula C7H5FN2
    Formula Weight 136.13
    MDL Number MFCD00015477
    UN Number UN3439
    Beilstein 3588945
    Synonym 6-Fluoroanthranilonitrile
    Chemical Name or Material 2-Amino-6-fluorobenzonitrile

    Specifications

    Formula Weight 136.13
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 77326-36-4
    Melting Point 125° to 129°C
    MDL Number MFCD00015477
    Quantity 5g
    Molecular Formula C7H5FN2
    UN Number UN3439
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Beilstein 3588945
    Synonym 6-Fluoroanthranilonitrile
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H311-H302-H332-H315-H319
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Chemical Name or Material 2-Amino-6-fluorobenzonitrile
    Assay 99%

    GHS H Statement
    H311-H302-H332-H315-H319
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit