Alfa Aesar™ 2-Amino-6-methoxy-3-nitropyridine, 97%

Manufacturer: Alfa Aesar™ H26668MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH26668MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 73896-36-3
    Molecular Formula C6H7N3O3
    Formula Weight 169.14
    MDL Number MFCD03206481
    Synonym 6-Methoxy-3-nitro-2-pyridinamine
    Chemical Name or Material 2-Amino-6-methoxy-3-nitropyridine

    Specifications

    Formula Weight 169.14
    Melting Point 170° to 172°C
    Synonym 6-Methoxy-3-nitro-2-pyridinamine
    Health Hazard 2 GHS H Statement
    H301-H311-H332-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C6H7N3O3
    MDL Number MFCD03206481
    CAS 73896-36-3
    Quantity 250mg
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2-Amino-6-methoxy-3-nitropyridine
    Assay 97%

    GHS H Statement
    H301-H311-H332-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit