Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-6-methoxybenzothiazole, 98%

Click to view available options
Quantity:
10g
50g
1747-60-0
C8H8N2OS
180.225
MFCD00005787
KZHGPDSVHSDCMX-UHFFFAOYSA-N
2-amino-6-methoxybenzothiazole,6-methoxybenzo d thiazol-2-amine,2-benzothiazolamine, 6-methoxy,6-methoxy-2-aminobenzothiazole,2-amino-6-methoxy benzothiazole,benzothiazole, 2-amino-6-methoxy,6-methoxy-2-benzothiazolamine,6-methoxybenzothiazol-2-ylamine,unii-lat2842yvx,ccris 1393
15630
6-methoxy-1,3-benzothiazol-2-amine
COC1=CC2=C(C=C1)N=C(S2)N
1747-60-0
C8H8N2OS
180.225
MFCD00005787
KZHGPDSVHSDCMX-UHFFFAOYSA-N
2-amino-6-methoxybenzothiazole,6-methoxybenzo d thiazol-2-amine,2-benzothiazolamine, 6-methoxy,6-methoxy-2-aminobenzothiazole,2-amino-6-methoxy benzothiazole,benzothiazole, 2-amino-6-methoxy,6-methoxy-2-benzothiazolamine,6-methoxybenzothiazol-2-ylamine,unii-lat2842yvx,ccris 1393
15630
6-methoxy-1,3-benzothiazol-2-amine
COC1=CC2=C(C=C1)N=C(S2)N
Melting Point 161°C to 164°C
Odor Odorless
Quantity 10g
Formula Weight 180.23
Percent Purity 98%
Chemical Name or Material 2-Amino-6-methoxybenzothiazole

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : DL2100000

TSCA : TSCA