Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-methoxybenzothiazole, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
250g
Packaging:
Glass bottle
1747-60-0
C8H8N2OS
180.23
MFCD00005787
KZHGPDSVHSDCMX-UHFFFAOYSA-N
2-amino-6-methoxybenzothiazole,6-methoxybenzo d thiazol-2-amine,2-benzothiazolamine, 6-methoxy,6-methoxy-2-aminobenzothiazole,2-amino-6-methoxy benzothiazole,benzothiazole, 2-amino-6-methoxy,6-methoxy-2-benzothiazolamine,6-methoxybenzothiazol-2-ylamine,unii-lat2842yvx,ccris 1393
15630
6-methoxy-1,3-benzothiazol-2-amine
COC1=CC2=C(C=C1)N=C(S2)N
1747-60-0
C8H8N2OS
180.23
MFCD00005787
KZHGPDSVHSDCMX-UHFFFAOYSA-N
2-amino-6-methoxybenzothiazole,6-methoxybenzo d thiazol-2-amine,2-benzothiazolamine, 6-methoxy,6-methoxy-2-aminobenzothiazole,2-amino-6-methoxy benzothiazole,benzothiazole, 2-amino-6-methoxy,6-methoxy-2-benzothiazolamine,6-methoxybenzothiazol-2-ylamine,unii-lat2842yvx,ccris 1393
15630
6-methoxy-1,3-benzothiazol-2-amine
COC1=CC2=C(C=C1)N=C(S2)N
Melting Point 162°C to 169°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Beige to White or Gray-Lilac
Quantity 250g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 27, II, 334
Formula Weight 180.23
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Amino-6-methoxybenzothiazole, 98%
Show More Show Less

GHS H Statement
Toxic if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.

GHS Signal Word: Danger

EINECSNumber : 217-130-0

RTECSNumber : DL2100000

TSCA : TSCA