Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-6-methoxypyridine, 97%

Click to view available options
Quantity:
5g
25g
17920-35-3
C6H8N2O
124.143
MFCD04972542
DEUALFRBMNMGDS-UHFFFAOYSA-N
2-amino-6-methoxypyridine,6-methoxy-pyridin-2-ylamine,6-methoxy-2-pyridinamine,6-methoxypyrid-2-ylamine,6-methoxy-2-pyridylamine,2-amino-6-methoxy pyridine,2-pyridinamine, 6-methoxy,methoxypyridin-6-amine,acmc-209efc,6-methoxy-2-aminopyridin
580053
6-methoxypyridin-2-amine
COC1=CC=CC(=N1)N
17920-35-3
C6H8N2O
124.143
MFCD04972542
DEUALFRBMNMGDS-UHFFFAOYSA-N
2-amino-6-methoxypyridine,6-methoxy-pyridin-2-ylamine,6-methoxy-2-pyridinamine,6-methoxypyrid-2-ylamine,6-methoxy-2-pyridylamine,2-amino-6-methoxy pyridine,2-pyridinamine, 6-methoxy,methoxypyridin-6-amine,acmc-209efc,6-methoxy-2-aminopyridin
580053
6-methoxypyridin-2-amine
COC1=CC=CC(=N1)N
Applications
It′s an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Sensitivity Air Sensitive
Color Brown
Quantity 5g
UN Number UN2810
Beilstein 115179
Solubility Information Slightly soluble in water.
Formula Weight 124.14
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Amino-6-methoxypyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon