Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-methylbenzoic acid, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
4389-50-8
C8H9NO2
151.165
XHYVBIXKORFHFM-UHFFFAOYSA-N
6-methylanthranilic acid,6-amino-o-toluic acid,benzoic acid, 2-amino-6-methyl,2-amino-6-methyl-benzoic acid,6-methylanthranilate,pubchem4957,acmc-209jvt,6-methyl anthranilic acid,3-amino-2-carboxytoluene,2-carboxy-3-methylaniline
151210
2-amino-6-methylbenzoic acid
CC1=C(C(=CC=C1)N)C(=O)O
4389-50-8
C8H9NO2
151.165
XHYVBIXKORFHFM-UHFFFAOYSA-N
6-methylanthranilic acid,6-amino-o-toluic acid,benzoic acid, 2-amino-6-methyl,2-amino-6-methyl-benzoic acid,6-methylanthranilate,pubchem4957,acmc-209jvt,6-methyl anthranilic acid,3-amino-2-carboxytoluene,2-carboxy-3-methylaniline
151210
2-amino-6-methylbenzoic acid
CC1=C(C(=CC=C1)N)C(=O)O
Quantity 5g
Color Beige to Tan
Infrared Spectrum Authentic
Melting Point 124°C to 129°C
Packaging Glass bottle
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula H2NC6H3(CH3)CO2H
Beilstein 14, I, 598
Formula Weight 151.16
Physical Form Powder, Crystals, and/or Lumps
Percent Purity ≥95%
Chemical Name or Material 2-Amino-6-methylbenzoic acid, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA