Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-6-methylheptane, 99%

Manufacturer:  Alfa Aesar™ A1201714

Quantity 25g

 View more versions of this product

Catalog No. AAA1201714

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

543-82-8
MFCD00008092
1209250
(±)-1,5-Dimethylhexylamine; (±)-6-Methyl-2-heptylamine
129.25
UN2733
14,6757
2-Amino-6-methylheptane

Specifications

543-82-8
0.767g/mL
Liquid
Amine-like
MFCD00008092
1209250
(±)-1,5-Dimethylhexylamine; (±)-6-Methyl-2-heptylamine
99%
1.4215
154° to 156°C
25g
129.25
UN2733
14,6757
2-Amino-6-methylheptane

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

RTECSNumber : MQ4840000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit