Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-6-methylpyridine, 99%

Manufacturer:  Alfa Aesar™ A1044436

Quantity 500g

 View more versions of this product

Catalog No. AAA1044436


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1824-81-3
108.14
UN2811
6-Amino-2-picoline; 6-Methyl-2-pyridinamine
C6H8N2
MFCD00006331
107048
2-Amino-6-methylpyridine

Specifications

1824-81-3
41° to 45°C
108.14
UN2811
107048
Unpleasant
2-Amino-6-methylpyridine
C6H8N2
208° to 209°C
MFCD00006331
500g
103°C (217°F)
6-Amino-2-picoline; 6-Methyl-2-pyridinamine
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H310-H315-H319-H335
Toxic if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P307+P311
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed: Call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : US1885000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit