Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-6-nitrobenzoic acid, 97%

Click to view available options
Quantity:
250mg
1g
5g
50573-74-5
C7H6N2O4
182.135
MFCD00034785
GGKYLHNARFFORH-UHFFFAOYSA-N
2-amino-6-nitrobenzoic acid,benzoic acid, 2-amino-6-nitro,6-nitroanthranilic acid,2-amino-6-nitro-benzoic acid,2-amino-6-nitrobenzoicacid,6-nitroanthanilic acid,acmc-209knw,6-nitro-anthranilic acid,6-nitro-2-aminobenzoesaure,ksc911o8d
95795
C1=CC(=C(C(=C1)[N+](=O)[O-])C(=O)O)N
50573-74-5
C7H6N2O4
182.135
MFCD00034785
GGKYLHNARFFORH-UHFFFAOYSA-N
2-amino-6-nitrobenzoic acid,benzoic acid, 2-amino-6-nitro,6-nitroanthranilic acid,2-amino-6-nitro-benzoic acid,2-amino-6-nitrobenzoicacid,6-nitroanthanilic acid,acmc-209knw,6-nitro-anthranilic acid,6-nitro-2-aminobenzoesaure,ksc911o8d
95795
C1=CC(=C(C(=C1)[N+](=O)[O-])C(=O)O)N
Quantity 1g
Beilstein 2807130
Formula Weight 182.13
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Amino-6-nitrobenzoic acid

storageNote1 : -30° to -10°C