Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-nitrobenzothiazole, 96%, Acros Organics

Click to view available options
Quantity:
25g
Packaging:
Plastic bottle
6285-57-0
C7H5N3O2S
195.2
MFCD00005786
GPNAVOJCQIEKQF-UHFFFAOYSA-N
2-amino-6-nitrobenzothiazole,6-nitrobenzo d thiazol-2-amine,2-benzothiazolamine, 6-nitro,6-nitrobenzothiazol-2-amine,6-nitro-2-aminobenzothiazole,2-amino-6-nitro benzothiazole,6-nitro-2-benzothiazolamine,benzothiazole, 2-amino-6-nitro,ccris 1391,6-nitro-benzothiazol-2-ylamine
22704
6-nitro-1,3-benzothiazol-2-amine
C1=CC2=C(C=C1[N+](=O)[O-])SC(=N2)N
6285-57-0
C7H5N3O2S
195.2
MFCD00005786
GPNAVOJCQIEKQF-UHFFFAOYSA-N
2-amino-6-nitrobenzothiazole,6-nitrobenzo d thiazol-2-amine,2-benzothiazolamine, 6-nitro,6-nitrobenzothiazol-2-amine,6-nitro-2-aminobenzothiazole,2-amino-6-nitro benzothiazole,6-nitro-2-benzothiazolamine,benzothiazole, 2-amino-6-nitro,ccris 1391,6-nitro-benzothiazol-2-ylamine
22704
6-nitro-1,3-benzothiazol-2-amine
C1=CC2=C(C=C1[N+](=O)[O-])SC(=N2)N
Melting Point 247°C to 252°C
Quantity 25g
Packaging Plastic bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 95% min. (HPLC)
Beilstein 27, II, 232
Formula Weight 195.2
Percent Purity 96%
Chemical Name or Material 2-Amino-6-nitrobenzothiazole, 97%

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-513-7