Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-(trifluoromethyl)benzoic acid, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
314-46-5
C8H6F3NO2
205.14
XZBQDBWIRHIZGJ-UHFFFAOYSA-N
2-amino-6-trifluoromethyl benzoic acid,benzoic acid, 2-amino-6-trifluoromethyl,6-trifluoromethyl anthranilic acid,pubchem4985,3-amino-2-carboxybenzotrifluoride,buttpark 24\07-93,6-trifluoromethyl anthranilicacid,2-carboxy-3-trifluoromethyl aniline,2-amino-6-trifluoromethyl benzoicacid
2778508
2-amino-6-(trifluoromethyl)benzoic acid
C1=CC(=C(C(=C1)N)C(=O)O)C(F)(F)F
314-46-5
C8H6F3NO2
205.14
XZBQDBWIRHIZGJ-UHFFFAOYSA-N
2-amino-6-trifluoromethyl benzoic acid,benzoic acid, 2-amino-6-trifluoromethyl,6-trifluoromethyl anthranilic acid,pubchem4985,3-amino-2-carboxybenzotrifluoride,buttpark 24\07-93,6-trifluoromethyl anthranilicacid,2-carboxy-3-trifluoromethyl aniline,2-amino-6-trifluoromethyl benzoicacid
2778508
2-amino-6-(trifluoromethyl)benzoic acid
C1=CC(=C(C(=C1)N)C(=O)O)C(F)(F)F
Packaging Glass Bottle
Melting Point 133.0°C to 136.0°C
Quantity 1g
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Formula Weight 205.14
Percent Purity 97%
Chemical Name or Material 2-Amino-6-(trifluoromethyl)benzoic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning