Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-9,9-dimethylfluorene, 98%

Click to view available options
Quantity:
250mg
1g
5g
108714-73-4
C15H15N
209.292
MFCD09953790
GUTJITRKAMCHSD-UHFFFAOYSA-N
2-amino-9,9-dimethylfluorene,9,9-dimethyl-9h-fluoren-2-amine,2-amino-9,9-dimethyl-9h-fluorene,9,9-dimethyl-9h-fluoren-2-ylamine,9,9-dimethyl9h-fluorene-2-amine,pubchem23234,ksc495c6d,9,9-dimethyl-2-aminofluorene,2-amino-9,9-dimethyl fluorene,9,9-dimethyl-fluoren-2-amine
22617093
9,9-dimethylfluoren-2-amine
CC1(C2=CC=CC=C2C3=C1C=C(C=C3)N)C
108714-73-4
C15H15N
209.292
MFCD09953790
GUTJITRKAMCHSD-UHFFFAOYSA-N
2-amino-9,9-dimethylfluorene,9,9-dimethyl-9h-fluoren-2-amine,2-amino-9,9-dimethyl-9h-fluorene,9,9-dimethyl-9h-fluoren-2-ylamine,9,9-dimethyl9h-fluorene-2-amine,pubchem23234,ksc495c6d,9,9-dimethyl-2-aminofluorene,2-amino-9,9-dimethyl fluorene,9,9-dimethyl-fluoren-2-amine
22617093
9,9-dimethylfluoren-2-amine
CC1(C2=CC=CC=C2C3=C1C=C(C=C3)N)C
Quantity 1g
Color White
Beilstein 15506865
Formula Weight 209.29
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2-Amino-9,9-dimethylfluorene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning