Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-p-cresol, 97%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
95-84-1
C7H9NO
123.16
MFCD00007699
ZMXYNJXDULEQCK-UHFFFAOYSA-N
2-amino-p-cresol,2-hydroxy-5-methylaniline,phenol, 2-amino-4-methyl,3-amino-4-hydroxytoluene,6-hydroxy-m-toluidine,4-methyl-2-aminophenol,5-methyl-2-hydroxyaniline,2-amino-4-cresol,p-cresol, 2-amino,2-amino-4-methyl-phenol
7264
2-amino-4-methylphenol
CC1=CC(=C(C=C1)O)N
95-84-1
C7H9NO
123.16
MFCD00007699
ZMXYNJXDULEQCK-UHFFFAOYSA-N
2-amino-p-cresol,2-hydroxy-5-methylaniline,phenol, 2-amino-4-methyl,3-amino-4-hydroxytoluene,6-hydroxy-m-toluidine,4-methyl-2-aminophenol,5-methyl-2-hydroxyaniline,2-amino-4-cresol,p-cresol, 2-amino,2-amino-4-methyl-phenol
7264
2-amino-4-methylphenol
CC1=CC(=C(C=C1)O)N
Melting Point 134.0°C to 139.0°C
Color Beige to Brown-Gray
Infrared Spectrum Authentic
Quantity 50g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula H2NC6H3(CH3)OH
Beilstein 13,601
Solubility Information Solubility in water: slightly soluble
Formula Weight 123.16
Physical Form Flakes or Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Amino-p-cresol, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-457-3

RTECSNumber : SJ6078000

TSCA : TSCA