Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoanthraquinone 80.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
117-79-3
C14H9NO2
223.231
MFCD00001233
XOGPDSATLSAZEK-UHFFFAOYSA-N
2-aminoanthraquinone,2-amino-9,10-anthraquinone,aminoanthraquinone,anthraquinone, 2-amino,9,10-anthracenedione, 2-amino,2-amino-9,10-anthracenedione,nsc 5,2-amino-anthraquinone,2-aminoanthra-9,10-quinone,beta-aminoanthraquinone
8341
CHEBI:34258
2-aminoanthracene-9,10-dione
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)N
117-79-3
C14H9NO2
223.231
MFCD00001233
XOGPDSATLSAZEK-UHFFFAOYSA-N
2-aminoanthraquinone,2-amino-9,10-anthraquinone,aminoanthraquinone,anthraquinone, 2-amino,9,10-anthracenedione, 2-amino,2-amino-9,10-anthracenedione,nsc 5,2-amino-anthraquinone,2-aminoanthra-9,10-quinone,beta-aminoanthraquinone
8341
CHEBI:34258
2-aminoanthracene-9,10-dione
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)N
Melting Point 302°C
Color Undesignated
Quantity 500g
Formula Weight 223.23
Physical Form Crystal-Powder at 20°C
Percent Purity ≥80.0% (HPLC)
Chemical Name or Material 2-Aminoanthraquinone

EINECSNumber : (4)-0706

RTECSNumber : CB5120000

TSCA : Yes