Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminobenzenesulfonamide, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
3306-62-5
C6H8N2O2S
172.202
MFCD00007932
YAZSBRQTAHVVGE-UHFFFAOYSA-N
orthanilamide,o-aminobenzenesulfonamide,o-sulfanilamide,benzenesulfonamide, 2-amino,2-amino-benzenesulfonamide,benzenesulfonamide, o-amino,2-aminobenzene-1-sulfonamide,o-aminobenzenesulphonamide,chembl6705,benzenesulfonamide, 2-amino-9ci
72894
2-aminobenzenesulfonamide
C1=CC=C(C(=C1)N)S(=O)(=O)N
3306-62-5
C6H8N2O2S
172.202
MFCD00007932
YAZSBRQTAHVVGE-UHFFFAOYSA-N
orthanilamide,o-aminobenzenesulfonamide,o-sulfanilamide,benzenesulfonamide, 2-amino,2-amino-benzenesulfonamide,benzenesulfonamide, o-amino,2-aminobenzene-1-sulfonamide,o-aminobenzenesulphonamide,chembl6705,benzenesulfonamide, 2-amino-9ci
72894
2-aminobenzenesulfonamide
C1=CC=C(C(=C1)N)S(=O)(=O)N
Melting Point 153°C to 157°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
Color Cream to White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula H2NC6H4SO2NH2
Formula Weight 172.21
Physical Form Crystalline Powder or Crystals
Percent Purity ≥97.5%
Chemical Name or Material 2-Aminobenzenesulfonamide
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.