Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminobenzenesulfonic Acid 99.0+%, TCI America™

Click to view available options
Quantity:
25g
100g
500g
88-21-1
C6H7NO3S
173.186
MFCD00007705
ZMCHBSMFKQYNKA-UHFFFAOYSA-N
orthanilic acid, aniline-2-sulfonic acid, o-aminobenzenesulfonic acid, o-sulfanilic acid, aniline-o-sulfonic acid, 2-sulfanilic acid, aniline-o-sulphonic acid, 1-aminobenzene-2-sulfonic acid, 2-aminobenzenesulphonic acid, benzenesulfonic acid, 2-amino
6926
CHEBI:1015
2-aminobenzenesulfonic acid
C1=CC=C(C(=C1)N)S(=O)(=O)O
88-21-1
C6H7NO3S
173.186
MFCD00007705
ZMCHBSMFKQYNKA-UHFFFAOYSA-N
orthanilic acid, aniline-2-sulfonic acid, o-aminobenzenesulfonic acid, o-sulfanilic acid, aniline-o-sulfonic acid, 2-sulfanilic acid, aniline-o-sulphonic acid, 1-aminobenzene-2-sulfonic acid, 2-aminobenzenesulphonic acid, benzenesulfonic acid, 2-amino
6926
CHEBI:1015
2-aminobenzenesulfonic acid
C1=CC=C(C(=C1)N)S(=O)(=O)O
Color White-Yellow
Quantity 100g
Formula Weight 173.19
Physical Form Crystal-Powder at 20°C
Percent Purity ≥99.0% (HPLC,T)
Chemical Name or Material 2-Aminobenzenesulfonic Acid

EINECSNumber : (3)-1971

RTECSNumber : DB4727700

TSCA : Yes