Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Aminobenzoyl-Ala-Gly-Leu-Ala-p-nitrobenzylamide

Click to view available options
Quantity:
0.5mg
1mg
100307-95-7
MFCD00237463
abz-ala-gly-leu-ala-p-nitrobenzylamide,2s-2-2-2s-2-2-aminophenyl formamido propanamido acetamido-4-methyl-n-1s-1-4-nitrophenyl methyl carbamoyl ethyl pentanamide,2-amino-n-2s-1-2-2s-4-methyl-1-2s-2-4-nitrophenyl methylamino propanoyl amino-1-oxopentan-2-yl amino-2-oxoethyl amino-1-oxopropan-2-yl benzamide
100307-95-7
MFCD00237463
abz-ala-gly-leu-ala-p-nitrobenzylamide,2s-2-2-2s-2-2-aminophenyl formamido propanamido acetamido-4-methyl-n-1s-1-4-nitrophenyl methyl carbamoyl ethyl pentanamide,2-amino-n-2s-1-2-2s-4-methyl-1-2s-2-4-nitrophenyl methylamino propanoyl amino-1-oxopentan-2-yl amino-2-oxoethyl amino-1-oxopropan-2-yl benzamide
Color White
Quantity 1mg
Solubility Information Soluble in water at 1mg/mL
Formula Weight 583.64
Physical Form Solid
Chemical Name or Material 2-Aminobenzoyl-Ala-Gly-Leu-Ala-p-nitrobenzylamide

storageNote1 : -30° to -10°C